N-Boc-(methylamino)acetaldehyde, 97%, Thermo Scientific Chemicals
N-Boc-(methylamino)acetaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-(methylamino)acetaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H50870.03
also known as H50870-03
Price (USD)/ Each
308.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
308.00
Add to cart
N-Boc-(methylamino)acetaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH50870.03
Price (USD)/ Each
308.00
-
Add to cart
Chemical Identifiers
CAS123387-72-4
IUPAC Nametert-butyl N-methyl-N-(2-oxoethyl)carbamate
Molecular FormulaC8H15NO3
InChI KeyMSWTVSDFEYSRMQ-UHFFFAOYSA-N
SMILESCN(CC=O)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay (HPLC)>97.5%
FormLiquid
N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-(methylamino)acetaldehyde is involved in the preparation of benzo[d]imidazole inhibitor of co-activator associated arginine methyltransferase.

Solubility
Slightly miscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Pandya, B. A.; Dandapani, S.; Duvall, J. R.; Rowley, A.; Mulrooney, C. A.; Ryba, T.; Dombrowski, M.; Harton, M.; Young, D. W.; Marcaurelle, L. A. Practical asymmetric synthesis of beta-hydroxy gamma-amino acids via complimentary aldol reactions. Tetrahedron 2011, 67 (34), 6131-6137.
  2. Wan, H.; Huynh, T.; Pang, S.; Geng, J.; Vaccaro, W.; Poss, M. A.; Trainor, G. L.; Lorenzi, M. V.; Gottardis, M.; Jayaraman, L.; Purandare, A. V. Benzo[d]imidazole inhibitors of coactivator associated arginine methyltransferase 1 (CARM1)-hit to lead studies. Bioorg. Med. Chem. Lett. 2009, 19 (17), 5063-5066.