Thermo Scientific Chemicals

Propionamidoxime, 97%, Thermo Scientific Chemicals

Catalog number: H50889.MD
250 mg, Each
Thermo Scientific Chemicals

Propionamidoxime, 97%, Thermo Scientific Chemicals

Catalog number: H50889.MD
250 mg, Each
Quantity
Catalog number: H50889.MD
also known as H50889-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29335-36-2
IUPAC Name
N'-hydroxypropanimidamide
Molecular Formula
C3H8N2O
InChI Key
RLZPCFQNZGINRP-UHFFFAOYSA-N
SMILES
CCC(N)=NO
Appearance (Color)
White to pale yellow
Assay (unspecified)
≥96.0%

Description

Propionamidoxime is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionamidoxime is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text