Thermo Scientific Chemicals

3-(4-Chlorophenoxy)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H50935.MD
250 mg, Each
Thermo Scientific Chemicals

3-(4-Chlorophenoxy)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H50935.MD
250 mg, Each
Quantity
Catalog number: H50935.MD
also known as H50935-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
384346-27-4
IUPAC Name
3-(4-chlorophenoxy)piperidine
Molecular Formula
C11H14ClNO
InChI Key
OXELOOCWZRBKSV-UHFFFAOYNA-N
SMILES
ClC1=CC=C(OC2CCCNC2)C=C1
Thin Layer Chromatography
96.0% min

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text