Thermo Scientific Chemicals

4-(3-Chlorophenoxy)piperidine, 98%, Thermo Scientific Chemicals

Catalog number: H50956.03
1 g, Each
Thermo Scientific Chemicals

4-(3-Chlorophenoxy)piperidine, 98%, Thermo Scientific Chemicals

Catalog number: H50956.03
1 g, Each
Quantity
Catalog number: H50956.03
also known as H50956-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97840-40-9
IUPAC Name
4-(3-chlorophenoxy)piperidine
Molecular Formula
C11H14ClNO
InChI Key
BYXWGMQMUJCYPK-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(OC2CCNCC2)=C1
Appearance (Color)
Clear colorless to yellow or orange
Assay (GC)
≥95.0%
Form
Viscous liquid
Refractive Index
1.5490-1.5510 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text