Thermo Scientific Chemicals

4-(3-Pyrrolidinyl)morpholine, 97%, Thermo Scientific Chemicals

Catalog number: H51002.03
1 g, Each
Thermo Scientific Chemicals

4-(3-Pyrrolidinyl)morpholine, 97%, Thermo Scientific Chemicals

Catalog number: H51002.03
1 g, Each
Quantity
Catalog number: H51002.03
also known as H51002-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53617-37-1
IUPAC Name
4-(pyrrolidin-3-yl)morpholine
Molecular Formula
C8H16N2O
InChI Key
OPJDRNMJJNFSNZ-UHFFFAOYNA-N
SMILES
C1CC(CN1)N1CCOCC1
Assay (GC)
96.0% min

Description

4-(3-Pyrrolidinyl)morpholine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(3-Pyrrolidinyl)morpholine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text