2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Kai Wang; F.Peter Guengerich. Bioactivation of fluorinated 2-aryl-benzothiazole antitumor molecules by human cytochrome P450s 1A1 and 2W1 and deactivation by cytochrome P450 2S1. Chem. Res. Toxicol. 2012, 25, (8),1740-1751
Ashley A.Weekes; Mark C.Bagley; Andrew D.Westwell. An efficient synthetic route to biologically relevant 2-phenylbenzothiazoles substituted on the benzothiazole ring. Tetrahedron. 2011, 67, (40),7743-7747