N-Boc-trans-4-hydroxy-L-prolinol, 96%, Thermo Scientific Chemicals
N-Boc-trans-4-hydroxy-L-prolinol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-trans-4-hydroxy-L-prolinol, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H51050.06
also known as H51050-06
Price (USD)/ Each
433.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
433.00
Add to cart
N-Boc-trans-4-hydroxy-L-prolinol, 96%, Thermo Scientific Chemicals
Catalog numberH51050.06
Price (USD)/ Each
433.00
-
Add to cart
Chemical Identifiers
CAS61478-26-0
IUPAC Nametert-butyl 4-hydroxy-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular FormulaC10H19NO4
InChI KeyUFJNFQNQLMGUTQ-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CC(O)CC1CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (unspecified)>95.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only