Thermo Scientific Chemicals

1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals

Catalog number: H51063.06
5 g, Each
Thermo Scientific Chemicals

1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals

Catalog number: H51063.06
5 g, Each
Quantity
Catalog number: H51063.06
also known as H51063-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23978-55-4
IUPAC Name
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium
Molecular Formula
C12H28N2O4
InChI Key
NLMDJJTUQPXZFG-UHFFFAOYSA-P
SMILES
C1COCCOCC[NH2+]CCOCCOCC[NH2+]1
Form
Powder or crystals or crystalline powder
Infrared spectrum
Conforms
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥95.0%

Description

1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text