1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Shayessteh Dadfarnia; Mojtaba Shamsipur. Highly selective membrane transport of Zn2+ ion by a cooperative carrier composed of 1,10-diaza-18-crown-6 and palmitic acid. Bulletin of the Chemical Society of Japan. 1992, 65, (10),2779-2783
Mojtaba Shamsipur; Alexander I.Popov. Multinuclear Nmr study of some alkali and thallium complexes with 1,10-diaza-18-crown-6 in nonaqueous solutions. Inorganica Chimica Acta. 1980, 43, 243-247