Thermo Scientific Chemicals

1-Aza-18-crown-6, 95%, Thermo Scientific Chemicals

Catalog number: H51065.03
1 g, Each
Thermo Scientific Chemicals

1-Aza-18-crown-6, 95%, Thermo Scientific Chemicals

Catalog number: H51065.03
1 g, Each
Quantity
Catalog number: H51065.03
also known as H51065-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33941-15-0
Appearance (Color)
White to yellow
Assay (unspecified)
>94.0%
Form
Crystalline powder

Description

1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aza-18-crown-6 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofunctionalized macrocycles. It is also used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text