Cyclopentanone-2,2,5,5-d{4}, 95%, Thermo Scientific Chemicals
Cyclopentanone-2,2,5,5-d{4}, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentanone-2,2,5,5-d{4}, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51095.06
also known as H51095-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclopentanone-2,2,5,5-d4
CAS3997-89-5
Health Hazard 1H226-H315-H319
Health Hazard 2GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only