Uracil-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
Uracil-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Uracil-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H51098.MD
also known as H51098-MD
Price (USD)/ Each
66.10
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
66.10
Add to cart
Uracil-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH51098.MD
Price (USD)/ Each
66.10
-
Add to cart
Specifications
Chemical Name or MaterialUracil-5-carboxaldehyde
CAS1195-08-0
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only