Thermo Scientific Chemicals

Cyclopropylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51158.AE
100 mL, Each
Thermo Scientific Chemicals

Cyclopropylmagnesium bromide, 0.5M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51158.AE
100 mL, Each
Quantity
Catalog number: H51158.AE
also known as H51158-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23719-80-4
IUPAC Name
bromo(cyclopropyl)magnesium
Molecular Formula
C3H5BrMg
InChI Key
QUPLLJGAJVXXBW-UHFFFAOYSA-M
SMILES
Br[Mg]C1CC1
Form
Liquid
Appearance (Color)
Yellow to yellow-brown to brown
Assay from Supplier's CofA
≥96.0%
Appearance (Color)
Yellow to yellow-brown to brown
Form
Liquid

Description

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text