Thermo Scientific Chemicals

3-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51161.AD
50 mL, Each
Thermo Scientific Chemicals

3-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51161.AD
50 mL, Each
Quantity
Catalog number: H51161.AD
also known as H51161-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
36229-42-2
IUPAC Name
bromo(3-chlorophenyl)magnesium
Molecular Formula
C6H4BrClMg
InChI Key
ZIYRUWPTUPFRQU-UHFFFAOYSA-M
SMILES
ClC1=CC([Mg]Br)=CC=C1
Concentration
0.80-1.20M
Form
Liquid

Description

3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1'-DiBoc-2,2'-Biaziridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorophenylmagnesium bromide is involved in the synthesis of enantiopure 2-imidazolidinones by reacting with 1,1′-DiBoc-2,2′-Biaziridine.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents. Reacts with water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text