Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.
Solubility
Reacts with water.
Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Kenneth N. Campbell; James O. Knobloch; Barbara K. Campbell. The Preparation and Some Reactions of 1,1,1-Trifluoro-2-alkenes.J. Am. Chem. Soc.,1950, 72(10), 4380-4384.
Rima S. Al-awar; Sajan P. Joseph; Daniel L. Comins. Conversion of N-acyl-2,3-dihydro-4-pyridones to 4-chloro-1,2-dihydropyridines using the Vilsmeier Reagent. Synthesis of (-)-coniine and (±)-lupinine. J. Org. Chem., 1993, 58(27), 7732-7739.