Thermo Scientific Chemicals

n-Propylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51162.AP
500 mL, Each
Thermo Scientific Chemicals

n-Propylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51162.AP
500 mL, Each
Quantity
Catalog number: H51162.AP
also known as H51162-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2234-82-4
IUPAC Name
chloro(propyl)magnesium
Molecular Formula
C3H7ClMg
InChI Key
AMXVKMXLDJXAPM-UHFFFAOYSA-M
SMILES
CCC[Mg]Cl
Form
Liquid
Comment
Solvent: MeTHF
Concentration
0.80-1.20M

Description

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). Also used as reactant in Grignard-reactions for the introduction of the n-propyl group.

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text