Thermo Scientific Chemicals

4-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51163.AE
100 mL, Each
Thermo Scientific Chemicals

4-Chlorophenylmagnesium bromide, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51163.AE
100 mL, Each
Quantity
Catalog number: H51163.AE
also known as H51163-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
873-77-8
IUPAC Name
bromo(4-chlorophenyl)magnesium
Molecular Formula
C6H4BrClMg
InChI Key
CDEMHJCJMMOFMB-UHFFFAOYSA-M
SMILES
ClC1=CC=C([Mg]Br)C=C1
Assay (Titration)
0.8-1.2 M
Appearance (Color)
Clear to slightly hazy
Form
Liquid

Description

4-Chlorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text