Thermo Scientific Chemicals

tert-Butylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51164.AE
100 mL, Each
Thermo Scientific Chemicals

tert-Butylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

Catalog number: H51164.AE
100 mL, Each
Quantity
Catalog number: H51164.AE
also known as H51164-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
677-22-5
IUPAC Name
tert-butyl(chloro)magnesium
Molecular Formula
C4H9ClMg
InChI Key
ZDRJSYVHDMFHSC-UHFFFAOYSA-M
SMILES
CC(C)(C)[Mg]Cl
Concentration
0.8-1.2M
Comment
Aqueous quench: passes test

Description

tert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.

Solubility
Miscible with alcohol and water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text