4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.
Solubility
Not miscible in water. Soluble in ether, hexane and tetrahydrofuran.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Bruce A.Reinhardt; Lawrence L.Brott; Stephen J.Clarson; Ann G.Dillard; Jayprakash C.Bhatt; Ramamurthi Kannan; Lixiang Yuan; Guang S.He; Paras N.Prasad. Highly active two-photon dyes: Design, synthesis, and characterization toward application. Chem. Mater. 1998, 10, (7),1863-1874