3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione, 97%, Thermo Scientific Chemicals
3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H51488.MD
also known as H51488-MD
Price (USD)/ Each
528.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
622.00 
Save 93.35 (15%)
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
528.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
622.00 
Save 93.35 (15%)
Add to cart
3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione, 97%, Thermo Scientific Chemicals
Catalog numberH51488.MD
Price (USD)/ Each
528.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
622.00 
Save 93.35 (15%)
-
Add to cart
Specifications
Chemical Name or Material3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione
CAS129034-70-4
Health Hazard 1H301-H312-H315-H319-H372
Health Hazard 2GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
View more
3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Store aw
RUO – Research Use Only

General References:

  1. Lanny S.Liebeskind; Richard W.Fengl. 3-Stannylcyclobutenediones as nucleophilic cyclobutenedione equivalents. Synthesis of substituted cyclobutenediones and cyclobutenedione monoacetals and the beneficial effect of catalytic copper iodide on the Stille reaction. J. Org. Chem. 1990, 55, (19),5359-5364 
  2. Lanny S.Liebeskind; Marvin S.Yu; Richard H.Yu; Jianying Wang; Karl S.Hagen. 4,4'-Bi(cyclobutene-1,2-diones): bisquaryls. J. Am. Chem. Soc. 1993, 115, (20),9048-9055