6-Hydroxypyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
6-Hydroxypyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Hydroxypyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H51673.03
also known as H51673-03
Price (USD)/ Each
266.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
266.00
Add to cart
6-Hydroxypyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog numberH51673.03
Price (USD)/ Each
266.00
-
Add to cart
Specifications
Chemical Name or Material6-Hydroxypyridine-3-boronic acid pinacol ester
CAS1054483-78-1
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC11H16BNO3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only