Thermo Scientific Chemicals

Ethyl 5-aminovalerate hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H51682.MD
250 mg, Each
Thermo Scientific Chemicals

Ethyl 5-aminovalerate hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H51682.MD
250 mg, Each
Quantity
Catalog number: H51682.MD
also known as H51682-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29840-57-1
IUPAC Name
ethyl 5-aminopentanoate hydrochloride
Molecular Formula
C7H16ClNO2
InChI Key
JELXJZASXLHKDV-UHFFFAOYSA-N
SMILES
Cl.CCOC(=O)CCCCN
Assay (unspecified)
95.0% min

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text