trans-4-Hydroxy-L-proline methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
trans-4-Hydroxy-L-proline methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-Hydroxy-L-proline methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H51726.14
also known as H51726-14
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
trans-4-Hydroxy-L-proline methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH51726.14
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS40216-83-9
IUPAC Namehydrogen methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate chloride
Molecular FormulaC6H12ClNO3
InChI KeyKLGSHNXEUZOKHH-JBUOLDKXSA-N
SMILES[H+].[Cl-].COC(=O)[C@@H]1C[C@@H](O)CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals
Assay (unspecified)>97.5%
Loss on Drying<0.50%
Optical purity>98.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only