4-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
4-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51745.03
also known as H51745-03
Price (USD)/ Each
91.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
91.70
Add to cart
4-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog numberH51745.03
Price (USD)/ Each
91.70
-
Add to cart
Specifications
Chemical Name or Material4-Ethynylbenzeneboronic acid pinacol ester
CAS1034287-04-1
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC14H17BO2
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only