Chemical Name or MaterialBenzyl thioacetimidate hydrochloride
CAS32894-07-8
Health Hazard 1H318
Health Hazard 2GHS H Statement H318 Causes serious eye damage.
Health Hazard 3P280-P305+P351+P338-P310
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.