Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals
Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H51844.06
also known as H51844-06
Price (USD)/ Each
94.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
94.90
Add to cart
Benzyl thioacetimidate hydrochloride, 96%, Thermo Scientific Chemicals
Catalog numberH51844.06
Price (USD)/ Each
94.90
-
Add to cart
Specifications
Chemical Name or MaterialBenzyl thioacetimidate hydrochloride
CAS32894-07-8
Health Hazard 1H318
Health Hazard 2GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3P280-P305+P351+P338-P310
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only