Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
100 mL
Catalog number H51864.AC
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
Add to cart
Tris(2-carboxyethyl)phosphine hydrochloride, 95%, 0.5M soln. in water, Thermo Scientific Chemicals
Catalog numberH51864.AC
Price (USD)/ Each
198.65
Online exclusive
220.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS51805-45-9
IUPAC Name3-[bis(2-carboxylatoethyl)phosphanyl]propanoate
Molecular FormulaC9H12O6P
InChI KeyPZBFGYYEXUXCOF-UHFFFAOYSA-K
SMILES[O-]C(=O)CCP(CCC([O-])=O)CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)0.45M - 0.55M
Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-carboxyethyl)phosphine hydrochloride is widely used as a reducing agent to reduce disulfide bonds in proteins and trans-4,5-dihydroxy-1,2-dithiane. It is involved in the reduction of sulfoxides, sulfonyl chlorides, N-oxides and azides. It plays an important role in the tissue homogenization process for RNA isolation. It acts as a chelating agent and forms complexes with various heavy metal ions such as Zn(II), Cd(II), Pb(II), and Ni(II). Furthermore, it is useful in the labeling of cysteine residues with maleimides.

Solubility
Miscible with water and methanol.

Notes
Air sensitive. Store in cool place. Incompatible with strong oxidizing agents
RUO – Research Use Only

General References:

  1. La, M.; Feng, Y.; Yang, C.; Chen, C. Performances of p-Aminophenol Redox Cycling by Thiols and Tris (2-carboxyethyl) phosphine on Cysteamine-and 3-Mercaptopropionic acid-Covered Gold Electrodes. Int. J. Electrochem. Sci. 2014, 9, 6985-6992.
  2. Miralles, G.; Verdié, P.; Puget, K.; Maurras, A.; Martinez, J.; Subra, G. Microwave-mediated reduction of disulfide bridges with supported (Tris (2-carboxyethyl) phosphine) as resin-bound reducing agent. ACS Comb. Sci. 2013, 15 (4), 169-173.