Thermo Scientific Chemicals
2-Chloro-4-fluoro-3-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Catalog number: H51879.03
1 g, Each
Thermo Scientific Chemicals
2-Chloro-4-fluoro-3-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Catalog number: H51879.03
1 g, Each
Quantity
Catalog number: H51879.03
also known as H51879-03
Price (USD)
Quantity
-
Have Questions?
Description
It is used in the synthesis and Antibacterial Evaluation of 6-Amino-8-methylquinolones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the synthesis and Antibacterial Evaluation of 6-Amino-8-methylquinolones.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.