Thermo Scientific Chemicals

4-(1-Piperidinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H51922.03
1 g, Each
Thermo Scientific Chemicals

4-(1-Piperidinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: H51922.03
1 g, Each
Quantity
Catalog number: H51922.03
also known as H51922-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
852227-96-4
IUPAC Name
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
Molecular Formula
C17H26BNO2
InChI Key
OTOKWHGMHAAFRM-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCCC1
Form
Solid
Purity
≥94.0%

Description

4-(1-Piperidinyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Piperidinyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text