Biphenyl-4,4'-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Biphenyl-4,4′-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Tao Tang; Tingting Lin; FuKe Wang; Chaobin He. Origin of near-infrared absorption for azulene-containing conjugated polymers upon protonation or oxidation. J. Phys. Chem. B 2015, 119, (25),8176-8183
- Tao Tang; Tingting Lin; FuKe Wang; Chaobin He. Azulene-based conjugated polymers with tuneable near-IR absorption up to 2.5 μm. Polym. Chem. 2014, 5, (8),2980-2989