Thermo Scientific Chemicals

N-Boc-4-iodo-L-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog number: H51960.14
25 g, Each
Thermo Scientific Chemicals

N-Boc-4-iodo-L-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog number: H51960.14
25 g, Each
Quantity
Catalog number: H51960.14
also known as H51960-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62129-44-6
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-iodophenyl)propanoic acid
Molecular Formula
C14H18INO4
InChI Key
JZLZDBGQWRBTHN-NSHDSACASA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(I)C=C1)C(O)=O
Assay (HPLC)
97.5% min

Description

N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-iodo-L-phenylalanine is used as pharmaceutical intermediate. It is also used as a p-iodo phenyl alanine derivative with N-Boc protection.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text