(S)-2-(Boc-amino)-4-phenylbutyric acid, 98%, Thermo Scientific Chemicals
(S)-2-(Boc-amino)-4-phenylbutyric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-2-(Boc-amino)-4-phenylbutyric acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51966.03
also known as H51966-03
Price (USD)/ Each
71.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
71.50
Add to cart
(S)-2-(Boc-amino)-4-phenylbutyric acid, 98%, Thermo Scientific Chemicals
Catalog numberH51966.03
Price (USD)/ Each
71.50
-
Add to cart
Chemical Identifiers
CAS100564-78-1
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-4-phenylbutanoic acid
Molecular FormulaC15H21NO4
InChI KeyMCODLPJUFHPVQP-LBPRGKRZSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)>97.5%
FormPowder
Optical Rotation+7.6? (C₁, EtOH)
(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-2-(Boc-amino)-4-phenylbutyric acid is used as organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Hong-Yan Shen; Gui-Ling Tian; Yun-Hua Ye; Jianbo Wang. Non-coded amino acids as acyl donor substrates for peptide bond formation catalyzed by thermoase in toluene. Journal of Molecular Catalysis B: Enzymatic. 2005, 37, (1-6),26-29
  2. Jean-Christophe Califano; Laurent Goullieux; Muriel Amblard; Jean Alain Fehrentz; Nicole Bernad; Gilbert Berge; Jean Castel; Jean Martinez. Synthesis and biological activities of peptidomimetic analogues of compound A71623, a potent and selective CCK-A receptor agonist. Letters in Peptide Science. 1997, 4, (4),235-239