N-Boc-4-bromo-L-phenylalanine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-4-bromo-L-phenylalanine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Ina Israel; Wolfgang Brandau; Georgios Farmakis; Samuel Samnick. Improved synthesis of no-carrier-added p-[124I]iodo-l-phenylalanine and p-[131I]iodo-l-phenylalanine for nuclear medicine applications in malignant gliomas. Applied Radiation and Isotopes. 2008, 66, (4),513-522
Syed T.Ahmed; Fabio Parmeggiani; Nicholas J.Weise; Sabine L.Flitsch; Nicholas J.Turner. Chemoenzymatic synthesis of optically pure l- and d-biarylalanines through biocatalytic asymmetric amination and palladium-catalyzed arylation. ACS Catal. 2015, 5, (9),5410-5413