Thermo Scientific Chemicals

4-Amino-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H51970.03
1 g, Each
Thermo Scientific Chemicals

4-Amino-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H51970.03
1 g, Each
Quantity
Catalog number: H51970.03
also known as H51970-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
943-80-6
IUPAC Name
(2S)-2-amino-3-(4-aminophenyl)propanoic acid
Molecular Formula
C9H12N2O2
InChI Key
CMUHFUGDYMFHEI-QMMMGPOBSA-N
SMILES
N[C@@H](CC1=CC=C(N)C=C1)C(O)=O
Optical Rotation
-42° ± 2° (c=2, water)
Appearance (Color)
White or cream to brown
Form
Crystals or powder or crystalline powder
Assay from Supplier's CofA
≥94.0%
Water Content (Karl Fischer Titration)
≤0.50%

Description

It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text