Thermo Scientific Chemicals

N-Fmoc-3-(2-naphthyl)-D-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H51974.03
1 g, Each
Thermo Scientific Chemicals

N-Fmoc-3-(2-naphthyl)-D-alanine, 95%, Thermo Scientific Chemicals

Catalog number: H51974.03
1 g, Each
Quantity
Catalog number: H51974.03
also known as H51974-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
138774-94-4
IUPAC Name
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-2-yl)propanoic acid
Molecular Formula
C28H23NO4
InChI Key
JYUTZJVERLGMQZ-AREMUKBSSA-N
SMILES
OC(=O)[C@@H](CC1=CC2=CC=CC=C2C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
White to pale cream
Form
Powder
Assay (HPLC)
≥94.0%
Identification (FTIR)
Conforms
Proton NMR
Conforms to structure

Description

N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text