N-Fmoc-L-beta-homoleucine, 95%, Thermo Scientific Chemicals
N-Fmoc-L-beta-homoleucine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-beta-homoleucine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H51976.MD
also known as H51976-MD
Price (USD)/ Each
132.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
132.00
Add to cart
N-Fmoc-L-beta-homoleucine, 95%, Thermo Scientific Chemicals
Catalog numberH51976.MD
Price (USD)/ Each
132.00
-
Add to cart
Chemical Identifiers
CAS193887-44-4
IUPAC Name(3S)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methylhexanoic acid
Molecular FormulaC22H25NO4
InChI KeyYLVSABQQLLRFIJ-HNNXBMFYSA-N
SMILESCC(C)C[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
FormPowder
Proton NMRConforms
Purity≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only