Thermo Scientific Chemicals

4-Iodo-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H51979.03
1 g, Each
Thermo Scientific Chemicals

4-Iodo-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H51979.03
1 g, Each
Quantity
Catalog number: H51979.03
also known as H51979-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24250-85-9
IUPAC Name
(2S)-2-amino-3-(4-iodophenyl)propanoic acid
Molecular Formula
C9H10INO2
InChI Key
PZNQZSRPDOEBMS-QMMMGPOBSA-N
SMILES
N[C@@H](CC1=CC=C(I)C=C1)C(O)=O
Assay (unspecified)
>94.0%

Description

4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.

Solubility
Partly miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text