N-Fmoc-L-pipecolic acid, 95%, Thermo Scientific Chemicals
N-Fmoc-L-pipecolic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-pipecolic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H51981.03
also known as H51981-03
Price (USD)/ Each
70.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
70.10
Add to cart
N-Fmoc-L-pipecolic acid, 95%, Thermo Scientific Chemicals
Catalog numberH51981.03
Price (USD)/ Each
70.10
-
Add to cart
Chemical Identifiers
CAS86069-86-5
IUPAC Name(2S)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidine-2-carboxylic acid
Molecular FormulaC21H21NO4
InChI KeyCKLAZLINARHOTG-IBGZPJMESA-N
SMILESOC(=O)[C@@H]1CCCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Infrared spectrumConforms
Appearance (Color)White
FormPowder
Mass SpecConforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only