Thermo Scientific Chemicals

4-Chloro-D-phenylalanine, 95%

Catalog number: H51982.03
1 g, Each
Thermo Scientific Chemicals

4-Chloro-D-phenylalanine, 95%

Catalog number: H51982.03
1 g, Each
Quantity
Catalog number: H51982.03
also known as H51982-03
Price (USD)
Quantity
-
Have Questions?

Description

It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text