Thermo Scientific Chemicals

(S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals

Catalog number: H51987.06
5 g, Each
Thermo Scientific Chemicals

(S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals

Catalog number: H51987.06
5 g, Each
Quantity
Catalog number: H51987.06
also known as H51987-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
162558-25-0
IUPAC Name
3-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular Formula
C23H26N2O6
InChI Key
PKAUMAVONPSDRW-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>94.0%

Description

As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text