N-Fmoc-3-(3-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
N-Fmoc-3-(3-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-3-(3-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H52056.06
also known as H52056-06
Price (USD)/ Each
594.65
Online exclusive
661.00 
Save 66.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
594.65
Online exclusive
661.00 
Save 66.35 (10%)
Add to cart
N-Fmoc-3-(3-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
Catalog numberH52056.06
Price (USD)/ Each
594.65
Online exclusive
661.00 
Save 66.35 (10%)
-
Add to cart
Chemical Identifiers
CAS175453-07-3
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-3-yl)propanoate
Molecular FormulaC23H19N2O4
InChI KeyJQLPMTXRCLXOJO-NRFANRHFSA-M
SMILES[O-]C(=O)[C@H](CC1=CC=CN=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0%
Appearance (Color)Powder or crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only