Thermo Scientific Chemicals

3,4-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H52057.03
1 g, Each
Thermo Scientific Chemicals

3,4-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H52057.03
1 g, Each
Quantity
Catalog number: H52057.03
also known as H52057-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
198560-43-9
IUPAC Name
(2S)-3-(3,4-difluorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular Formula
C24H19F2NO4
InChI Key
IHSYIDJNVXPQRM-QFIPXVFZSA-N
SMILES
OC(=O)[C@H](CC1=CC=C(F)C(F)=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Proton NMR
Conforms
Optical Rotation
|b = -42? ?1? (C=1 in DMF)
Solution Test
Clear solution (0.3 g in 2mL DMF)
Water Content (Karl Fischer Titration)
≤2.0%
Appearance (Color)
White to pale cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text