N-Boc-3-nitro-L-phenylalanine, 95%, Thermo Scientific Chemicals
N-Boc-3-nitro-L-phenylalanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-3-nitro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H52058.06
also known as H52058-06
Price (USD)/ Each
574.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
574.00
Add to cart
N-Boc-3-nitro-L-phenylalanine, 95%, Thermo Scientific Chemicals
Catalog numberH52058.06
Price (USD)/ Each
574.00
-
Add to cart
Chemical Identifiers
CAS131980-29-5
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-nitrophenyl)propanoic acid
Molecular FormulaC14H18N2O6
InChI KeyOWTGPXDXLMNQKK-NSHDSACASA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=CC(=C1)[N+]([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥94.0%
Chiral LC≥99% (ee)
Water Content (Karl Fischer Titration)≤0.5%
Proton NMRConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only