3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals
3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H52069.06
also known as H52069-06
Price (USD)/ Each
959.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
959.00
Add to cart
3,5-Difluoro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals
Catalog numberH52069.06
Price (USD)/ Each
959.00
-
Add to cart
Specifications
Chemical Name or Material3,5-Difluoro-N-Fmoc-L-phenylalanine
CAS205526-24-5
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC24H19F2NO4
View more
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-N-Fmoc-L-phenylalanine is used for producing high quality peptides.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only