4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Pradhan, D.; Priyadarshini, V.; Munikumar, M.; Swargam, S.; Umamaheswari, A.; Bitla, A. Para-(benzoyl)-phenylalanine as a potential inhibitor against LpxC of Leptospira spp.: homology modeling, docking, and molecular dynamics study. J. Biomol. Struct. Dyn. 2014, 32 (2), 171-185.
Liu, W.; Alfonta, L.; Mack, A. V.; Schultz, P. G. Structural Basis for the Recognition of para-Benzoyl-L-phenylalanine by Evolved Aminoacyl-tRNA Synthetases. Angew. Chem. Int. Ed. 2007, 46 (32), 6073-6075.
Dorman, G.; Olszewski, J. D.; Prestwich, G. D.; Hong, Y.; Ahern, D. G. Synthesis of Highly Tritiated 4-Benzoyl-L-phenylalanine, a Photoactivatable Amino Acid. J. Org. Chem. 1995, 60 (7), 2292-2297.