Nalpha-Fmoc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine, 95%, Thermo Scientific Chemicals
Nalpha-Fmoc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nalpha-Fmoc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52115.MD
also known as H52115-MD
Price (USD)/ Each
208.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
208.00
Add to cart
Nalpha-Fmoc-N^w-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine, 95%, Thermo Scientific Chemicals
Catalog numberH52115.MD
Price (USD)/ Each
208.00
-
Add to cart
Specifications
Chemical Name or MaterialNalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-beta-homoarginine
CAS401915-53-5
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC35H42N4O7S
View more
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only