Thermo Scientific Chemicals

4-Trifluoromethyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H52170.MD
250 mg, Each
Thermo Scientific Chemicals

4-Trifluoromethyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H52170.MD
250 mg, Each
Quantity
Catalog number: H52170.MD
also known as H52170-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
114926-38-4
IUPAC Name
2-amino-3-[4-(trifluoromethyl)phenyl]propanoic acid
Molecular Formula
C10H10F3NO2
InChI Key
CRFFPDBJLGAGQL-UHFFFAOYNA-N
SMILES
NC(CC1=CC=C(C=C1)C(F)(F)F)C(O)=O
LCMS
Conforms
Purity
≥94.0%
Form
Solid

Description

4-fluoro-l-phenylalanine (fF), or 4-trifluoromethyl-l-phenylalanine were introduced inside the chain of peptides to enhances their antimicrobial activity..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-fluoro-l-phenylalanine (fF), or 4-trifluoromethyl-l-phenylalanine were introduced inside the chain of peptides to enhances their antimicrobial activity..

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text