N-Fmoc-3-(4-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
N-Fmoc-3-(4-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-3-(4-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52172.MD
also known as H52172-MD
Price (USD)/ Each
71.70
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
71.70
Add to cart
N-Fmoc-3-(4-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals
Catalog numberH52172.MD
Price (USD)/ Each
71.70
-
Add to cart
Specifications
Chemical Name or MaterialN-Fmoc-3-(4-pyridyl)-L-alanine
CAS169555-95-7
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC23H20N2O4
View more
Fmoc-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fmoc-∫-(4-pyridyl)-Ala-OH is potentially useful for solid phase peptide synthesis techniques.

Solubility
Soluble in 0.1g in 4mL DMF(clearly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only
Vommina V. Sureshbabu,; N. Narendra and G. Nagendra. Chiral N-Fmoc-β-Amino Alkyl Isonitriles Derived from Amino Acids: First Synthesis and Application in 1-Substituted Tetrazole Synthesis. J. Org. Chem., 2009, 74 (1), 153-157