(S)-3-(Boc-amino)-5-methylhexanoic acid, 95%, Thermo Scientific Chemicals
(S)-3-(Boc-amino)-5-methylhexanoic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-3-(Boc-amino)-5-methylhexanoic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H52173.03
also known as H52173-03
Price (USD)/ Each
384.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
384.00
Add to cart
(S)-3-(Boc-amino)-5-methylhexanoic acid, 95%, Thermo Scientific Chemicals
Catalog numberH52173.03
Price (USD)/ Each
384.00
-
Add to cart
Specifications
Chemical Name or Material(S)-3-(Boc-amino)-5-methylhexanoic acid
CAS132549-43-0
Melting Point53°C to 55°C
Recommended StorageAmbient temperatures
TSCANo
View more
(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of carba peptides and building block for β-peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-(Boc-amino)-5-methylhexanoic acid is used as building blocks for the synthesis of “carba peptides” and building block for β-peptides.

Solubility
Very slightly soluble (1.1 g/L at 25°C).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Daniel H.Rich; Byung Jo Moon; Amrit S.Boparai. Synthesis of (2S,3R)-3-amino-2-hydroxy-5-methylhexanoic acid derivatives. Application to the synthesis of amastatin, an inhibitor of aminopeptidases. J. Org. Chem. 1980, 45, (12),2288-2290