Chemical Identifiers
CAS201484-50-6
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3,3-diphenylpropanoic acid
Molecular FormulaC30H25NO4
InChI KeyPENQOTJCVODUQU-NDEPHWFRSA-N
SMILESOC(=O)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(C1=CC=CC=C1)C1=CC=CC=C1
View more